Learn More
Tocris Bioscience™ Hematoxylin
Histologic stain; used to stain cell nuclei
Marca: Tocris Bioscience™ 5222/25G
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Hematoxylin is a dye used to stain cell nuclei. Hematoxylin is commonly combined with eosin for H&E histological staining and is a component of the Papanicolaou stain used in cytology. Hematoxylin can be used with other fixatives to stain muscle striations, fibrin, myelin and elastin. Can also be used for H&E staining of intact tissues.Specifica
| Histology Stains | |
| 517-28-2 | |
| 25 g |
| (6 AS,11 bR)-7,11 b-Dihydrobenz[b]indeno[1,2-d]pyran-3,4,6 A,9,10(6 H)-pentol | |
| Room Temperature |
Titolo del prodotto
Individuate un'opportunità di miglioramento?
For Research Use Only