Descrizione
- Accessory
- Filter classification H14 (acc. to. EN 1822-1:2009, >99.995% at 0.3 μm)
- Not in conjunction with reinforced heating output, reinforced fan
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | M 56 E3.1 Drying/Heating Oven |
| Materiale | Plastic |
| Tipo | HEPA Filter |