missing translation for 'onlineSavingsMsg'
Learn More

HEPES, 0.5M buffer soln., pH 9.0, Thermo Scientific™

Catalog Number p-7042744
Click to view available options
Quantità:
100 mL
250 mL
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
7365-45-9
C8H18N2O4S
238.30
MFCD00006158
JKMHFZQWWAIEOD-UHFFFAOYSA-N
23831
CHEBI:42334
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
OCCN1CCN(CCS(O)(=O)=O)CC1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
7365-45-9
C8H18N2O4S
238.30
MFCD00006158
JKMHFZQWWAIEOD-UHFFFAOYSA-N
23831
CHEBI:42334
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
OCCN1CCN(CCS(O)(=O)=O)CC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico HEPES buffer soln.
Concentrazione 0.5 M
pH 9.0
Forma fisica Liquid
Quantità 100 mL
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'