missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hephaestin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Specifications
Antigene | Hephaestin |
---|---|
Concentrazione | 0.17 mg/mL |
Applicazioni | Immunohistochemistry (Paraffin) |
Classificazione | Polyclonal |
Coniugato | Unconjugated |
Product Code | Brand | Quantità | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantità | Price | Quantity & Availability | |||||
16808033
|
Proteintech
11148-1-AP-20UL |
20 μL |
140.00€
20µL |
Spedizione stimata: 08-05-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
16898023
|
Proteintech
11148-1-AP-150UL |
150 μL |
399.00€
150µL |
Spedizione stimata: 08-05-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Description
B1AJX8(HEPH) is similar to an iron transport protein found in mouse. The mouse protein is similar to ceruloplasmin, a serum multi-copper ferroxidase, and is thought to be a membrane-bound protein responsible for transport of dietary iron from epithelial cells of the intestinal lumen into the circulatory system. In mouse, defects in this gene can lead to severe microcytic anemia. Three transcript variants encoding different isoforms have been described for this gene. The protein encoded by this gene is similar to an iron transport protein found in mouse. The mouse protein is similar to ceruloplasmin, a serum multi-copper ferroxidase, and is thought to be a membrane-bound protein responsible for transport of dietary iron from epithelial cells of the intestinal lumen into the circulatory system. In mouse, defects in this gene can lead to severe microcytic anemia. Two transcript variants encoding different isoforms have been described for this gene.Specifications
Hephaestin | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q920H8, Q9BQS7, Q9Z0Z4 | |
117240, 15203, 9843 | |
Hephaestin Fusion Protein Ag1604 | |
Primary | |
-20°C | |
HEPH |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CPL, HEPH, Hephaestin, KIAA0698 | |
HEPH | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |