missing translation for 'onlineSavingsMsg'
Learn More

HEPPSO, 0.2M buffer soln., pH 8.0, Thermo Scientific™

Catalog Number p-7058224
Click to view available options
Quantità:
100 mL
250 mL
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
68399-78-0
C9H20N2O5S
268.328
MFCD00038351
GIZQLVPDAOBAFN-UHFFFAOYSA-N
100205
CHEBI:32951
2-hydroxy-3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
C1CN(CCN1CCO)CC(CS(=O)(=O)O)O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
68399-78-0
C9H20N2O5S
268.328
MFCD00038351
GIZQLVPDAOBAFN-UHFFFAOYSA-N
100205
CHEBI:32951
2-hydroxy-3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
C1CN(CCN1CCO)CC(CS(=O)(=O)O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico HEPPSO buffer soln.
Concentrazione 0.2 M
pH 8.0
Forma fisica Liquid
Quantità 100 mL
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'