missing translation for 'onlineSavingsMsg'
Learn More

Heptanoic acid, 98+%, Thermo Scientific Chemicals

Catalog Number p-7023798
25.66 EUR valido fino al 2024-03-29
Usa il codice promo "21615" per avere il tuo prezzo promozionale.


Click to view available options
Quantità:
500 mL
2500 mL
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
111-14-8
C7H14O2
130.187
MFCD00004426
MNWFXJYAOYHMED-UHFFFAOYSA-N
enanthic acid, oenanthic acid, n-heptanoic acid, enanthylic acid, heptoic acid, n-heptoic acid, n-heptylic acid, heptylic acid, oenanthylic acid, 1-hexanecarboxylic acid
8094
CHEBI:45571
heptanoic acid
CCCCCCC(=O)O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
111-14-8
C7H14O2
130.187
MFCD00004426
MNWFXJYAOYHMED-UHFFFAOYSA-N
enanthic acid, oenanthic acid, n-heptanoic acid, enanthylic acid, heptoic acid, n-heptoic acid, n-heptylic acid, heptylic acid, oenanthylic acid, 1-hexanecarboxylic acid
8094
CHEBI:45571
heptanoic acid
CCCCCCC(=O)O

Heptanoic acid is used as an organic building block for the synthesis of a variety of chemical compounds. It is involved in the esterification of steroids and used in the preparation of active pharmaceutical ingredients such as trenbolone enanthate, testosterone enanthate, drostanolone enanthate and methenolone enanthate. It is also used as an internal standard during gas chromatography analysis of the butyl esters of volatile acids. Its ester derivatives are used in the fragrance and flavor industries as well as cosmetics and industrial lubricants in aviation, refrigeration and automobile. It is used to prepare sodium heptanoate, which is used as a corrosion inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Heptanoic acid is used as an organic building block for the synthesis of a variety of chemical compounds. It is involved in the esterification of steroids and used in the preparation of active pharmaceutical ingredients such as trenbolone enanthate, testosterone enanthate, drostanolone enanthate and methenolone enanthate. It is also used as an internal standard during gas chromatography analysis of the butyl esters of volatile acids. Its ester derivatives are used in the fragrance and flavor industries as well as cosmetics and industrial lubricants in aviation, refrigeration and automobile. It is used to prepare sodium heptanoate, which is used as a corrosion inhibitor.

Solubility
Miscible with acetone, ethanol, diethyl ether, dimethylformamide, dimethyl sulfoxide and nitric acid. Slightly soluble in carbon tetrachloride and water.

Notes
Incompatible with strong oxidizing agents, bases and reducing agents.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico Heptanoic acid
Punto di fusione -8°C
Densità 0.917
Punti di ebollizione 222°C to 223°C
Punto d'infiammabilità 114°C (237°F)
Formula lineare CH3(CH2)5CO2H
Indice di rifrazione 1.423
Quantità 500 mL
Numero UN UN3265
Beilstein 1744723
Indice di Merck 14,4660
Informazioni di solubilità Miscible with acetone,ethanol,diethyl ether,dimethylformamide,dimethyl sulfoxide and nitric acid. Slightly soluble in carbon tetrachloride and water.
Peso formulazione 130.19
Percent Purity ≥98%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'