Learn More
Hexadecyltrimethylammonium Bromide-d9, TRC
Specifica
Specifica
| Materiale o nome chimico | Hexadecyltrimethylammonium-d9 Bromide |
| CAS | 95217-14-4 |
| Formula molecolare | C19 2H9 H33 N . Br |
| Quantità | 50 mg |
| Sinonimo | 1-Hexadecanaminium, N,N,N-tri(methyl-d3)-, bromide (1:1), 1-Hexadecanaminium, N,N,N-tri(methyl-d3)-, bromide (9CI), Cetrimonium-D9 Bromide (N,N,N-tri(methyl-D3)) |
| InChI Formula | InChI=1S/C19H42N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;/h5-19H2,1-4H3;1H/q+1;/p-1/i2D3,3D3,4D3; |
| SMILES | [Br-].[2H]C([2H])([2H])[N+](CCCCCCCCCCCCCCCC)(C([2H])([2H])[2H])C([2H])([2H])[2H] |
| IUPAC Name | hexadecyl-tris(trideuteriomethyl)azanium;bromide |
| Molecular Weight (g/mol) | 373.5 |
| Peso formulazione | 372.3066 |