missing translation for 'onlineSavingsMsg'
Learn More

Hexaethyl phosphorous triamide, 97%, Thermo Scientific Chemicals

Catalog Number p-7020508
Click to view available options
Quantità:
5 g
25 g
100 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
2283-11-6
MFCD00009041
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
2283-11-6
MFCD00009041

Hexaethyl phosphorous triamide is used in the Barton syntheses of hindered alkenes, involving desulfurization of small-ring heterocycles with elimination of nitrogen or carbon dioxide. It is also used as a reagent for selective vicinal dehalogenation of halogenated alkanes to give perhalogenated alkenes. The method has potential for the synthesis of highly fluorinated and mixed fluoro-halo alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexaethyl phosphorous triamide is used in the Barton syntheses of hindered alkenes, involving desulfurization of small-ring heterocycles with elimination of nitrogen or carbon dioxide. It is also used as a reagent for selective vicinal dehalogenation of halogenated alkanes to give perhalogenated alkenes. The method has potential for the synthesis of highly fluorinated and mixed fluoro-halo alkenes.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico Hexaethyl phosphorous triamide
Densità 0.903
Punti di ebollizione 245°C to 246°C
Punto d'infiammabilità 58°C (136°F)
Formula molecolare C8H4F3NO
Indice di rifrazione 1.4737
Quantità 5 g
Numero UN UN1993
Beilstein 636187
Sensibilità Air Sensitive
Sinonimo 2-trifluoromethyl phenyl isocyanate, 1-isocyanato-2-trifluoromethyl benzene, benzene, 1-isocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isocyanate, o-trifluoromethylphenylisocyanate, isocyanic acid, alpha,alpha,alpha-trifluoro-o-tolyl ester, o-trifluoromethyl phenyl isocyanate, 2-trifluoromethyl phenylisocyanate, 2-trifluoromethyl benzenisocyanate, isocyanic acid, .alpha.,.alpha.,.alpha.-trifluoro-o-tolyl ester
Informazioni di solubilità Insoluble in water.
InChI Key GZWGTVZRRFPVAS-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)C(F)(F)F)N=C=O
IUPAC Name 1-isocyanato-2-(trifluoromethyl)benzene
Molecular Weight (g/mol) 187.12
PubChem CID 16794
Peso formulazione 247.37
Percent Purity 97%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'