Learn More
Hexamethonium bromide, 98+%
Product Code. 15443239
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15443239 100 g 100g
15433239 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15443239 Supplier Thermo Scientific Alfa Aesar Supplier No. J60466.22
 Restricted Item
Spedizione stimata: 15-12-2026
Add to basket
Nicotinic acetyl choline receptor antagonist | CAS: 55-97-0 | C12H30Br2N2 | 362.194 g/mol
Nicotinic acetyl choline receptor antagonist
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 55-97-0
Molecular Formula C12H30Br2N2
Molecular Weight (g/mol) 362.194
MDL Number MFCD00011787
InChI Key FAPSXSAPXXJTOU-UHFFFAOYSA-L
Synonym hexamethonium bromide, hexamethonium dibromide, simpatoblock, gangliostat, esametina, hexameton, vegolysen, vegolysin, bistrium bromide, hexonium dibromide
PubChem CID 5938
IUPAC Name trimethyl-[6-(trimethylazaniumyl)hexyl]azanium;dibromide
SMILES C[N+](C)(C)CCCCCC[N+](C)(C)C.[Br-].[Br-]
Specifications
Melting Point 282°C to 285°C (decomposition)
Color White
Linear Formula (CH3)3N(Br)(CH2)6N(Br)(CH3)3
Quantity 100 g
Beilstein 3715749
Merck Index 14,4688
Formula Weight 362.19
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Hexamethonium bromide