Learn More
Hexarelin Acetate, TRC
Product Code. 30134978
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134978 50 mg 50mg
30132919 250 mg 250mg
30008665 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134978 Supplier Toronto Research Chemicals Supplier No. TRCH29470050MG
 Restricted Item
Spedizione stimata: 07-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 208251-52-9
Synonym L-Histidyl-2-methyl-D-tryptophyl-L-alanyl-L-tryptophyl-D-phenylalanyl-L-lysinamide Acetate∣Examorelin Acetate;
Specifications
Purity >95% (HPLC)
Format Neat
Molecular Weight (g/mol) 887
Quantity 50 mg
Shipping Condition Room Temperature