Learn More
CPC Colder Products Company™ HFC Series Non-Valved Coupling Connectors
Manufactured from polypropylene. CPC Colder Products Company™ HFC Series Non-Valved Coupling Connectors are manufactured from polypropylene. The series are ergonomically designed and easy to grip and simple to operate.
Marca:  CPC Colder Products Company™ 24835
Dettagli aggiuntivi : Peso : 0.80000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 11953794
Quantità Prezzo
1 48.50€ / Pezzo
10 43.38€ / Pezzo
Spedizione stimata
27-05-2021
Aggiungi al carrello
Allerta:
WARNING: Pressure, temperature, chemicals and operating environment can affect the performance of couplings. It is the customer’s responsibility to test the suitability of Colder products in their own application conditions.
Descrizione
Descrizione
  • High-efficiency valve
  • Ergonomic design
  • Polypropylene material
  • Compatible
  • More flow in a compact size
  • Easy to grip, simple to operate
Specifica
Specifica
Gray
60psi, 4.2 bar
0°C
71°C
Polypropylene