missing translation for 'onlineSavingsMsg'
Learn More

HIF-1 alpha Antibody (H1alpha 67-7), DyLight 350, Novus Biologicals™

Codice prodotto. 18022776 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.1 mL
Dimensione della confezione:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18022776 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18022776 Fornitore Novus Biologicals N. del fornitore NB100130UV

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody

HIF-1 alpha Monoclonal specifically detects HIF-1 alpha in Human, Monkey samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifica

Antigene HIF-1 alpha
Applicazioni Western Blot
Classificazione Monoclonal
Clone H1alpha 67-7
Coniugato DyLight 350
Diluizione Western Blot
Formulazione 50 mM sodium borate with 0.05% sodium azide
Alias gene AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp
Simboli geni HIF1A
Specie ospite Mouse
Immunogeno This HIF-1 alpha Antibody (H1alpha 67-7) was developed against a fusion protein containing amino acids 432 - 528 of human HIF-1 alpha [Uniprot# Q16665].
Peso molecolare dell'antigene 93 kDa
Metodo di purificazione Protein G purified
Quantità 0.1 mL
Disciplina di ricerca Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators
Primario o secondario Primary
ID gene (immissione) 3091.0
Target Species Human, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.