Description
- Intensity: 2000 μW/cm2
- Soft rubber viewport provides a comfortable viewing area with adequate space for eyeglasses
- Block out ambient light with lightweight access curtain
- Protect your eyes with contrast control filter
- Removable bottom panel allows cabinet to be placed over large samples or a transilluminator
- Lightweight access curtain blocks out ambient light
- Model features four longwave UV tubes mounted on one side of the cabinet
Specifications
Specifications
| Wattage | 4 x 15 W |
| Wavelength | 365 nm |
| Dimensioni (L x P x A) | 23.375 x 15.75 x 13 in. |
| Requisiti elettrici | 4 x 15 W, 230 V, 50 Hz |
| Product Type | UV Viewing Cabinet |