Learn More
Chemetrics™ High Range COD Photometer
Product Code. 11752103
Change view
Click to view available options
For Use With (Equipment):
CHEMetrics 0-1500 ppm HR COD vials (Mfr. Nos. K-7360S, K-7365, K-7361S, K-7366) and 0-15,000 ppm HR+ COD vials (Mfr. Nos. K-7370S, K-7375, K-7371S, K-7376)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11752103 CHEMetrics 0-1500 ppm HR COD vials (Mfr. Nos. K-7360S, K-7365, K-7361S, K-7366) and 0-15,000 ppm HR+ COD vials (Mfr. Nos. K-7370S, K-7375, K-7371S, K-7376) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11752103 Supplier Chemetrics™ Supplier No. A7325
  • ONLINE EXCLUSIVE PRICE
    652.00€
    609.12€ / pezzi 1
    Risparmia 42.88€
    6% Sconto
 Restricted Item
Spedizione stimata: 11-12-2026
Specifications
For Use With (Equipment) CHEMetrics 0-1500 ppm HR COD vials (Mfr. Nos. K-7360S, K-7365, K-7361S, K-7366) and 0-15,000 ppm HR+ COD vials (Mfr. Nos. K-7370S, K-7375, K-7371S, K-7376)
Includes 4 AAA batteries
Type COD Photometer (high range)