Learn More
Emma Safety Footwear Himalaya Safety Shoes
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
49
Dimensione della confezione:
1 paio
Descrizione
- S3-Safety shoe with sole plate for wet work
- Oil Nubuck construction
- GripForce™ Easy Twist
- 3M™ Reflective Striping
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifica
Specifica
| Certificazione/Conformità | EN ISO 20345:2011 |
| Tipo di chiusura | Laces |
| Colore | Black |
| Progettazione | Himalaya |
| Materiale rivestimento interno | Hydro-Tec™ Sanitized Silver |
| Materiale | Oiled Nubuck |
| Size | 35 |
| Materiale dello zoccolo | Steel |