Learn More
Histone Deacetylase 2/HDAC2 Antibody - BSA Free, Novus Biologicals™
Codice prodotto. 18632887 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.025 mg
0.1 mg
Dimensione della confezione:
0.03mg
0.10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18632887 0.025 mg 0.03mg
18190649 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 18632887 Fornitore Novus Biologicals N. del fornitore NBP2412850.025mg
Please to purchase this item. Need a web account? Register with us today!
Rabbit Polyclonal Antibody
Histone Deacetylase 2/HDAC2 Polyclonal specifically detects Histone Deacetylase 2/HDAC2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifica
Antigene Histone Deacetylase 2/HDAC2
Applicazioni Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classificazione Polyclonal
Concentrazione 1 mg/mL
Coniugato Unconjugated
Diluizione Western Blot 0.5-1 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence 20 μg/mL
Formulazione PBS with 0.02% Sodium Azide
Alias gene EC 3.5.1.98, HD2, histone deacetylase 2, RPD3, transcriptional regulator homolog RPD3, YAF1, YY1-associated factor 1
Simboli geni HDAC2
Specie ospite Rabbit
Immunogeno Antibody was raised against an 18 amino acid peptide near the carboxy terminus of human HDAC2. The immunogen is located within the last 50 amino acids of HDAC2. Amino Acid Squence: KKARIEEDKKETEDKKTD
Peso molecolare dell'antigene 52 kDa
Metodo di purificazione Affinity Purified
Quantità 0.025 mg
Status giuridico RUO
Disciplina di ricerca Breast Cancer, Cancer, Chromatin Research, Epigenetics, Histone Deacetylases, Stem Cell Markers
Primario o secondario Primary
ID gene (immissione) 3066
Test di specificità At least two isoforms of HDAC2 are known to exist; this antibody will detect both isoforms. HDAC2 antibody is predicted to not cross-react with other members of the HDAC family.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG