missing translation for 'onlineSavingsMsg'
Learn More

Histone H1.0 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Codice prodotto. 16857414 Sfoglia Tutto Proteintech Prodotti
Cambia vista
Click to view available options
Quantità:
20 μL
150 μL
Dimensione della confezione:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
16857414 20 μL 20µL
16847414 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 16857414 Fornitore Proteintech N. del fornitore 175101AP20UL

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Polyclonal Antibody

PTEN was identified as a tumor suppressor that is mutated in a large number of cancers at high frequency. The PTEN gene is a phosphatidylinositol-3, 4, 5-trisphosphate 3-phosphatase. It contains a tensin like domain as well as a catalytic domain similar to that of the dual specificity protein tyrosine phosphatases. Unlike most of the protein tyrosine phosphatases, this protein preferentially dephosphorylates phosphoinositide substrates. PTEN negatively regulates intracellular levels of phosphatidylinositol-3, 4, 5-trisphosphate in cells and functions as a tumor suppressor by negatively regulating AKT/PKB signaling pathway. PTEN plays a role as a key modulator of the AKT-mTOR signaling pathway controlling the tempo of the process of newborn neurons integration during adult neurogenesis, including correct neuron positioning, dendritic development and synapse formation. Also, PTEN may be a negative regulator of insulin signaling and glucose metabolism in adipose tissue. The nuclear monoubiquitinated form possesses greater apoptotic potential, whereas the cytoplasmic nonubiquitinated form of PTEN induces less tumor suppressive ability.

Specifica

Antigene Histone H1.0
Applicazioni Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry
Classificazione Polyclonal
Concentrazione 0.21 mg/mL
Coniugato Unconjugated
Formulazione PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene H1F0
N. accesso geni P07305, P10922, P43278
Alias gene H1 histone family, member 0, H10, H1F0, H1FV, Histone H1, Histone H1(0), Histone H1.0
Simboli geni H1f0
Specie ospite Rabbit
Immunogeno Histone H1.0 Fusion Protein Ag9982
Metodo di purificazione Antigen Affinity Chromatography
Quantità 20 μL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 14958, 24437, 3005
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Forma Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.