missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIV1 gp120 Polyclonal Antibody, HRP
Description
The PA1-73097 antibody reacts with HIV-1 gp120. PA1-73097 has been successfully used in ELISA, IHC, and IB applications. The PA1-73097 antibody was raised against Purified native gp120 from strain IIIB. NOTE: Sodium azide is a potent inhibitor of peroxidase!.
Human immunodeficiency virus type 1 (HIV-1) entry into target cells is directed by the envelope (Env) glycoproteins, which are present on the surface of HIV-1 virion or infected cells in the form of trimers consisting of gp120/gp41 complexes. The surface subunit, gp120, initiates the entry process by interacting sequentially with the CD4 receptor and a co-receptor CCR5 or CXCR4, thereby inducing a conformational change that allows the transmembrane (TM) gp41 subunit to mediate fusion between viral and target cell membranes. Cleavage of Env into its gp120 and gp41 components is necessary for activation of its fusogenic activity.
Specifications
Specifications
| Antigene | HIV1 gp120 |
| Applicazioni | ELISA, Immunohistochemistry, Western Blot |
| Classificazione | Polyclonal |
| Concentrazione | 1 mg/mL |
| Coniugato | HRP |
| Formulazione | PBS with 10mg/mL BSA and 0.002% thimerosal; pH 7.2 |
| Alias gene | HIV Glycoprotein 120; HIV-1 Glycoprotein 120; Human immunodeficiency virus |
| Specie ospite | Goat |
| Immunogeno | Purified native gp120 from strain IIIB. |
| Metodo di purificazione | IgG fraction |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?