Learn More
HLA DPA Antibody (TAL3C3), Alexa Fluor™ 488, Novus Biologicals™
Codice prodotto. 18776280 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.1 mL
Dimensione della confezione:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18776280 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18776280 Fornitore Novus Biologicals N. del fornitore NBP250601AF488
Mouse Monoclonal Antibody
HLA DPA Monoclonal antibody specifically detects HLA DPA in Human samples. It is validated for Western Blot, Immunohistochemistry, Radioimmune Assays
TRUSTED_SUSTAINABILITY
Specifica
Antigene HLA DPA
Applicazioni Western Blot, Immunohistochemistry, Radioimmune Assays (RIA)
Classificazione Monoclonal
Clone TAL3C3
Coniugato Alexa Fluor 488
Formulazione 50 mM Sodium Borate
Alias gene DP alpha 1 chain, HLA class II histocompatibility antigen, DP alpha chain, HLADP, HLA-SB alpha chain, major histocompatibility complex, class II, DP alpha 1, MHC class II antigen, MHC class II DP3-alpha, MHC class II DPA1, MHC class II HLA-DPA1 antigen, Primed lymphocyte test-1
Specie ospite Mouse
Immunogeno Bristol 8 separated alpha chain preparation
Metodo di purificazione Protein A purified
Quantità 0.1 mL
Status giuridico RUO
Disciplina di ricerca Adaptive Immunity, Asthma, Immunology
Primario o secondario Primary
ID gene (immissione) 3113
Target Species Human
Content And Storage Store at 4°C in the dark.
Forma Purified
Isotype IgG1