missing translation for 'onlineSavingsMsg'
Learn More

HLA-DR Antibody (IMMU-114) [Janelia Fluor™ 669], Novus Biologicals™

Codice prodotto. 30178396
Cambia vista
Click to view available options
Quantità:
0.1 mL
Dimensione della confezione:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30178396 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30178396 Fornitore Novus Biologicals (Bio-Techne) N. del fornitore NBP328647JF669

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Human Monoclonal Antibody

HLA-DR Monoclonal antibody specifically detects HLA-DR in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifica

Antigene HLA-DR
Applicazioni Flow Cytometry, ELISA, Functional Assay
Classificazione Monoclonal
Clone IMMU-114
Coniugato Janelia Fluor 669
Formulazione 50mM Sodium Borate
Alias gene FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW
Specie ospite Human
Immunogeno HLA-DR
Metodo di purificazione Protein A purified
Quantità 0.1 mL
Status giuridico RUO
Disciplina di ricerca Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cellular Markers, Diabetes Research, Immunology, Major Histocompatibility Complexes
Primario o secondario Primary
ID gene (immissione) 3122
Target Species Human
Content And Storage Store at 4°C in the dark.
Forma Purified
Isotype IgG4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.