missing translation for 'onlineSavingsMsg'
Learn More

HMGB1/HMG-1 Antibody - BSA Free, Novus Biologicals™

Codice prodotto. 18710204 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.025 mg
0.1 mg
Dimensione della confezione:
0.03mg
0.10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18710204 0.1 mg 0.10mg
18659697 0.025 mg 0.03mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18710204 Fornitore Novus Biologicals N. del fornitore NBP241264

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Polyclonal Antibody

HMGB1/HMG-1 Polyclonal specifically detects HMGB1/HMG-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Spécification

Antigene HMGB1/HMG-1
Applicazioni Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classificazione Polyclonal
Concentrazione 1 mg/mL
Coniugato Unconjugated
Diluizione Western Blot 1 ug/mL, ELISA, Immunohistochemistry 1-2 ug/mL, Immunocytochemistry/Immunofluorescence 20ug/mL, Immunohistochemistry-Paraffin
Formulazione PBS with 0.02% Sodium Azide
Alias gene Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein
Simboli geni HMGB1
Specie ospite Rabbit
Immunogeno Antibody was raised against a 19 amino acid peptide near the center of human HMGB1. The immunogen is located within amino acids 70 - 120 of HMGB1. Amino Acid Squence: KGETKKKFKDPNAPKRPPS
Peso molecolare dell'antigene 24 kDa
Metodo di purificazione Affinity Purified
Quantità 0.1 mg
Status giuridico RUO
Disciplina di ricerca Cellular Markers, Chromatin Research, Histones and Modified Histones, Neuroscience, Transcription Factors and Regulators
Primario o secondario Primary
ID gene (immissione) 3146
Test di specificità HMGB1 antibody is human, mouse and rat reactive.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.