missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HMOX1 Polyclonal Antibody

Descrizione
Recommended positive controls: HeLa, Raw264.7, C2C12, rat spleen, HeLa (500 μM CoCl2 treatment for 24 hr). Predicted reactivity: Mouse (81%), Rat (80%), Dog (85%), Pig (82%), Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Heme oxygenase (HO) is a microsomal enzyme that catalyzes the oxidation of heme to the antioxidant molecules, biliverdin and carbon monoxide. HO consists of two homologous isozymes, an inducible HO-1 and a constitutively expressed HO-2. HO-1 is induced by a wide variety of stimuli including conditions of oxidative stress, inflammatory agents, transforming growth factor beta and heat shock. The increase in expression of HO-1 is thought to be a cellular defense mechanism against oxidative stress since elevated HO could eventually generate more bilirubin, an anti-oxidant.
Specifica
Specifica
| Antigene | HMOX1 |
| Applicazioni | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classificazione | Polyclonal |
| Concentrazione | 1.55 mg/mL |
| Coniugato | Unconjugated |
| Formulazione | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | HMOX1 |
| N. accesso geni | P06762, P09601, P14901 |
| Alias gene | bK286B10; D8Wsu38e; heat shock protein 32; heat shock protein, 32-kD; heme orygenase 1; heme oxygenase; heme oxygenase (decycling) 1; heme oxygenase (decyclizing) 1; heme oxygenase 1; heme oxygenase-1; Hemox; hemoxygenase; Heox; HEOXG; Hmox; Hmox1; HMOX1D; HO; Ho1; HO-1; Hsp32; P32 protein |
| Simboli geni | HMOX1 |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?