Learn More
Sperian™ Schermo facciale Honeywell™ Clearways
Description
- Recommended for the agriculture, chemical and forestry industries
- Protects against hazards including abrasion, airborne particles, biohazards, chemicals, impacts, splashes and more
- Safe, practical, economical integral visor fitting ensures extra protection
Order Info
Note: The image shows both the Browguard and the Visor. Both pieces must be ordered separately. See Browguard (11886980), and Visors (11806990, 12652733, or 11896980).
Specifications
Specifications
| Lens Coating | Uncoated |
| Product Type | Schermo facciale Clearways |
| Modello | CV83P |
| N. per confezione | 1 ciasc. |
| Lens Material | Policarbonato |