Learn More
Scientific Industries SI™ Horizontal Tube Holder
Product Code. 15517355 Shop All Ohaus Products
Change view
Click to view available options
Holds:
12 x 15 mL Tubes
24 Microtubes
50 mL Tubes
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
15517355 12 x 15 mL Tubes Each
15507355 50 mL Tubes Each
15527355 24 Microtubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15517355 Supplier Scientific Industries SI™ Supplier No. 83041275
  • ONLINE EXCLUSIVE PRICE
    147.74€
    106.81€ / pezzi 1
    Risparmia 40.93€
    27% Sconto
View all promotions
In stock
Add to basket
Edge
Add to basket
Snap tubes into place swiftly and easily. Scientific Instruments SI™ Horizontal Tube Holder does not require additional attachments for use.
  • Holds tubes in place horizontally.
  • Holder snaps on to any Vortex-Genie™ 2 mixer and the Vortex-Genie™ Pulse
  • No additional attachments are necessary
  • Holds tubes in place horizontally.
  • Holder snaps on to any Vortex-Genie™ 2 mixer and the Vortex-Genie™ Pulse
  • No additional attachments are necessary
TRUSTED_SUSTAINABILITY
Specifications
Holds 12 x 15 mL Tubes
For Use With (Equipment) Vortex-Genie 2 Mixer and the Vortex-Genie Pulse
No. of Positions 12