Learn More
Sperian™ Howard Leight™ Multi Max™ Uncorded Disposable Earplugs
Codice prodotto. 11817030
Cambia vista
Click to view available options
Color: Yellow and Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto.
11817030 Color: Yellow and Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11817030 Fornitore Sperian™ N. del fornitore 3301109
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Denne vare kan ikke returneres. Se returpolicy
Offers flexibility, protection and economy in hearing protection with it two-sizes-in-one design, improving individual fit while simplifying inventory. Sperian™ Howard Leight™ Multi Max™ Uncorded Disposable Earplugs have a comfortable fit with self-adjusting polyurethane foam.
  • Smooth, soil-resistant closed cell foam skin prevents dirt build-up
  • Self-adjusting polyurethane foam expands to fit virtually every wearer
  • Ideal for agriculture and farming, airport ground crews, building construction and many others
TRUSTED_SUSTAINABILITY
Tekniske data
Colore Yellow, Red
Quantità 200/Pk.
Plug Shape Pill
Product Type Earplug
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.