Learn More
Invitrogen™ HOXD9 Monoclonal Antibody (OTI1H3)
GREENER_CHOICE
Codice prodotto. 15956784
Cambia vista
Click to view available options
Quantità:
100 μL
Dimensione della confezione:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
15956784 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 15956784 Fornitore Invitrogen™ N. del fornitore MA527194
Mouse Monoclonal Antibody
HOXD9 Monoclonal Antibody for Western Blot, IHC (P)
HOXD9 is the sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located at 2q31-2q37 chromosome regions. Deletions that removed the entire HOXD gene cluster or 5' end of this cluster have been associated with severe limb and genital abnormalities. The exact role of this gene has not been determined.
TRUSTED_SUSTAINABILITY
Specifica
Antigene HOXD9
Applicazioni Immunohistochemistry (Paraffin), Western Blot
Classificazione Monoclonal
Clone OTI1H3
Concentrazione 1 mg/mL
Coniugato Unconjugated
Formulazione PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HOXD9
N. accesso geni P28356
Alias gene homeo box D9; homeobox D9; homeobox Hox-D9; homeobox protein Hox-4.4; Homeobox protein Hox-4C; homeobox protein Hox-5.2; Homeobox protein Hox-D9; HOX4; Hox-4.3; Hox-4.3, mouse, homolog of; Hox-4.4; HOX4C; Hox-5.2; HOXD9; Hoxd-9
Simboli geni HOXD9
Specie ospite Mouse
Immunogeno Human recombinant protein fragment corresponding to amino acids 109-352 of HOXD9 produced in E.coli.
Metodo di purificazione Affinity Chromatography
Quantità 100 μL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 3235
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Forma Liquid
Isotype IgG1