missing translation for 'onlineSavingsMsg'
Learn More

IDEXX Water HPC for Quanti-Tray

IDEXX Water HPC for Quanti-Tray is an accurate test for determining heterotrophic plate counts in a 100ml sample.

Marca:  IDEXX Water 98-27390-00

Dettagli aggiuntivi : Peso : 0.61800kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15951190

  • dietro preventivo / 200 tests

Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.

Richiedi qui il prezzo per questo articolo.


Descrizione

Descrizione

  • Heterotrophic plate count made easy for bottling and manufacturing plants and on-premise testing
  • Quantifies heterotrophic organisms in a 100 mL format
  • No autoclaving, filtering, or agar
  • No colony counting or interpretation
  • Convenient 100-mL sample size—test source water, transport vehicle water, manufacturing line water, and finished products
  • Simple procedure needs no special training
  • Shares the same Quanti-Tray system as Colilert, Enterolert, and Pseudalert tests for maximum efficiency
  • Results valid at 44–72 hours
Specifica

Specifica

Pink
200/Pk.
Store at 2 to 30°C, away from light
44 hr.
Quanti-Tray
100 mL
Quantification
Heterotrophic plate count
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali