Learn More
Justrite™ HPLC Poly Manifold Fittings for Quick-Disconnect Disposal Cans
Use with manfiolds for Justrite™ Centura Quick-Disconnect HPLC Safety Disposal Cans
Marca: Justrite™ 28189
Dettagli aggiuntivi : Peso : 0.09000kg
Specifica
Quick-Disconnect Disposal Can | |
US OSHA, NFPA Code 30 | |
0.125 in. | |
HPLC poly manifold, 0.125 in. O.D., 0.062 in. I.D. |
Resin | |
0.3 cm | |
6 connectors | |
Each |