Learn More
IKA™ HS 260 Basic Shaker
Codice prodotto. 10142241 Sfoglia Tutto IKA Prodotti
Cambia vista
Click to view available options
Display:
LED Line Display
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Display Dimensione della confezione
10142241 LED Line Display Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 10142241 Fornitore IKA™ N. del fornitore 0003066600
  • ONLINE EXCLUSIVE PRICE
    3125.00€
    2921.66€ / pezzi 1
    Risparmia 203.34€
    6% Sconto
Visualizza tutte le promozioni
Spedizione stimata: 22-12-2026
Use this compact, flat reciprocal shaker for applications with a shaking weight of up to 7.5kg. IKA™ HS 260 Basic Shaker features a wide range of attachment combinations which makes it possible to use almost all shapes and sizes of vessels.
  • HS 260 basic and KS 260 basic
  • Electronic adjustment of speed and time with LED display
  • Wide range of attachment combinations makes it possible to use almost all shapes and sizes of vessels
  • Attachments are not included, please order separatelyHS 260 control and KS 260 controlSame features as HS/KS 260 basic, but in addition:
  • Digital display makes it possible to read the speed, timer function and operating modes
  • RS 232 interface
  • All functions can be controlled and documented with labworldsoft™
  • Electronic time switching clock: 0 to 9h 59min or continuous operation (∞)
  • With integrated end point positioning (for automated robot-controlled sampling)
TRUSTED_SUSTAINABILITY
Specifica
Intervallo di velocità 0 to 3000 rpm
Requisiti elettrici 45 W
Tensione 220/240, 100/120
Temperature Range (Metric) 5°C to 50°C
Certificazione/Conformità DIN EN 60529
Frequenza 50/60 Hz
Profondità (sistema metrico) 420 mm
Altezza (sistema metrico) 100 mm
Larghezza (sistema metrico) 360 mm
Modello HS 260 Basic
Display LED Line Display
Stile della piattaforma Flat
Intervallo temporizzatore 0 to 9:59 hr., Continuous