missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPA1L Mouse anti-Human, Mouse, Rat, Clone: 3E7H2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66780-1-IG-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This antibody well recognizes the endogenous HSPA1L protein in multiple cell lines.
In response to adverse changes in their environment, cells from many organisms increase the expression of a class of proteins referred to as heat shock proteins. One class of stress proteins, termed the "small heat shock proteins" is comprised of a diverse group of proteins from ∽15 to >30 kDa, with a single form found in yeast and human cells and multiple related forms found in higher plants. A direct relationship between elevated stress protein expression and neoplasia has been suggested from studies on estrogen-induced proteins in breast cancer cells. In addition, Hsp27 exhibits rapid increased phosphorylation in response to various mitogens, tumor promoter (e. g. phorbol esters) and calcium ionophores. The expression of a 24 kDa protein was shown to correlate with steroid hormone receptor in cell lines, organs, and tumors. The 24 kDa estrogen-induced protein was found to be expressed at high levels in carcinoma of the breast. Another study has suggested that levels of the Hsp27 correlate with endometrial adenocarcinomas. Interestingly, the regulation of human Hsp27 by estrogens and heat shock is reminiscent of the ecdysteroid and heat shock activation of the drosophila Hsp20 genes.Specifica
HSPA1L | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P16627, P34931, P55063 | |
Hspa1l | |
HSPA1L Fusion Protein Ag5182 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b |
Western Blot | |
3E7H2 | |
Unconjugated | |
HSPA1L | |
Heat shock 70 kDa protein 1L, HSP70 1L, HSP70 HOM, HSP70T, HSPA1L, hum70t | |
Mouse | |
Protein A | |
RUO | |
15482, 24963, 3305 | |
-20°C | |
Liquid |