Learn More
Abnova™ Human AATK Partial ORF (AAH47378, 161 a.a. - 260 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009625-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The protein encoded by this gene contains a tyrosine kinase domain at the N-terminus and a proline-rich domain at the C-terminus. This gene is induced during apoptosis, and expression of this gene may be a necessary pre-requisite for the induction of growth arrest and/or apoptosis of myeloid precursor cells. This gene has been shown to produce neuronal differentiation in a neuroblastoma cell line. [provided by RefSeq]
Sequence: SPEFVLKEAQEGCEPQAFAELASEGEGPGPETRLSTSLSGLNEKNPYRDSAYFSDLEAEAEATSGPEKKCGGDRAPGPELGLRSTGQPSEQVCLRPGVSGSpecifica
AAH47378 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SPEFVLKEAQEGCEPQAFAELASEGEGPGPETRLSTSLSGLNEKNPYRDSAYFSDLEAEAEATSGPEKKCGGDRAPGPELGLRSTGQPSEQVCLRPGVSG | |
RUO | |
AATK | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9625 | |
AATK (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AATYK/AATYK1/KIAA0641/LMR1/LMTK1/p35BP | |
AATK | |
Recombinant | |
wheat germ expression system |