Learn More
Abnova™ Human ABCF2 Full-length ORF (NP_009120.1, 1 a.a. - 623 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00010061-P01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intracellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, and White). This protein is a member of the GCN20 subfamily. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq]
Sequence: MPSDLAKKKAAKKKEAAKARQRPRKGHEENGDVVTEPQVAEKNEANGRETTEVDLLTKELEDFEMKKAAARAVTGVLASHPNSTDVHIINLSLTFHGQELLSDTKLELNSGRRYGLIGLNGIGKSMLLSAIGKREVPIPEHIDIYHLTREMPPSDKTPLHCVMEVDTERAMLEKEAERLAHEDAECEKLMELYERLEELDADKAEMRASRILHGLGFTPAMQRKKLKDFSGGWRMRVALARALFIRPFMLLLDEPTNHLDLDACVWLEEELKTFKRILVLVSHSQDFLNGVCTNIIHMHNKKLKYYTGNYDQYVKTRLELEENQMKRFHWEQDQIAHMKNYIARFGHGSAKLARQAQSKEKTLQKMMASGLTERVVSDKTLSFYFPPCGKIPPPVIMVQNVSFKYTKDGPCIYNNLEFGIDLDTRVALVGPNGAGKSTLLKLLTGELLPTDGMIRKHSHVKIGRYHQHLQEQLDLDLSPLEYMMKCYPEIKEKEEMRKIIGRYGLTGKQQVSPIRNLSDGQKCRVCLAWLAWQNPHMLFLDEPTNHLDIETIDALADAINEFEGGMMLVSHDFRLIQQVAQEIWVCEKQTITKWPGDILAYKEHLKSKLVDEEPQLTKRTHNVSpecifica
NP_009120.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
97.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ABCF2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10061 | |
ABCF2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ABC28/DKFZp586K1823/EST133090/HUSSY-18/HUSSY18/M-ABC1 | |
ABCF2 | |
Recombinant | |
wheat germ expression system |