Learn More
R&D Systems™ Human ACE/CD143 (NP_000780.1) VersaClone cDNA
Descrizione
Inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
Specifica
Specifica
| Gene | ACE/CD143 |
| Numero di accesso | NP_000780.1 |
| Concentrazione | 10 μg at 0.2 μg/ μL |
| Formulazione | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Tipo di inserto | 3934 bp |
| ID gene (immissione) | 1636 |
| Alias gene | ACE1angiotensin converting enzyme, somatic isoform, angiotensin I converting enzyme (peptidyl-dipeptidase A) 1, carboxycathepsin, CD143, CD143 antigen, DCP, DCP1, DCP1angiotensin-converting enzyme, dipeptidyl carboxypeptidase 1, Dipeptidyl carboxypeptidase I, EC 3.2.1.-, EC 3.4.15.1, Kininase II, MGC26566, MVCD3, peptidase P, testicular ECA |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Specie | Human |
| Quantità | 10 μg |