Learn More
Abcam Human ACE2 knockout Caco-2 cell line
Codice prodotto. 32140480
Cambia vista
Click to view available options
1000000 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto.
32140480 1000000 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 32140480 Fornitore Abcam N. del fornitore ab2737311000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 05-10-2026
Aggiungi al carrello
Aggiungi al carrello
missing translation for 'validMainframeMsg'
ACE2 KO cell line available to order. KO validated by Immunocytochemistry, Western blot. Knockout achieved by using CRISPR/Cas9, Homozygous: 62 bp deletion in exon 2.
ACE2 KO cell line available to order. KO validated by Immunocytochemistry, Western blot. Knockout achieved by using CRISPR/Cas9, Homozygous: 62 bp deletion in exon 2.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Quantità 1,000,000 Cells
Status giuridico RUO
Titolo del prodotto
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.