missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Activin RIIA (NP_001607) VersaClone cDNA

Codice prodotto. 18779517
Cambia vista
Click to view available options
Quantità:
10 μg
Dimensione della confezione:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18779517 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18779517 Fornitore R&D Systems™ N. del fornitore RDC1345

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Contains gene inserts that are fully sequenced and produced with high quality

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

The Human Activin RIIA (NP_001607) VersaClone cDNA clone contains the complete ORF for Activin RIIA, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Activin RIIA insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature

TRUSTED_SUSTAINABILITY

Specifica

Gene Activin RIIA
Numero di accesso NP_001607
Concentrazione 10 μg at 0.2 μg/μL
Formulazione cDNA is provided in 10mM Tris-Cl, pH 8.18
Tipo di inserto 1555bp
ID gene (immissione) 92
Alias gene activin A receptor, type II, activin A receptor, type IIA, Activin receptor type IIA, activin receptor type-2A, ActivinRIIA, ACTRII, ACTRIIA, ACVR2A, ACVR2ACTR-IIA, AVR2A, EC 2.7.11, EC 2.7.11.30
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Specie Human
Quantità 10 μg

For Reasearch Use Only. Not for use in Humans.

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.