Learn More
Abnova™ Human ALOXE3 Full-length ORF (AAI01939.1, 1 a.a. - 711 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
dietro preventivo
Specifica
Nome | ALOXE3 (Human) Recombinant Protein (P01) |
---|---|
Recombinant | Yes |
Metodo di purificazione | Glutathione Sepharose 4 Fast Flow |
Peso molecolare | 106.9kDa |
Requisiti di stoccaggio | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Descrizione
This gene is a member of the lipoxygenase family, which are catabolized by arachidonic acid-derived compounds. The encoded enzyme is a hydroperoxide isomerase that synthesizes a unique type of epoxy alcohol (8R-hydroxy-11R,12R-epoxyeicosa-5Z,9E,14Z-trienoic acid) from 12R-hydroperoxyeicosatetraenoic acid (12R-HPETE). This epoxy alcohol can activate the the nuclear receptor peroxisome proliferator-activated receptor alpha (PPARalpha), which is implicated in epidermal differentiation. Loss of function of the enzyme encoded by this gene results in ichthyosis, implicating the function of this gene in the differentiation of human skin. This gene is part of a cluster of lipoxygenase genes on 17p13.1. Mutations in this gene result in nonbullous congenital ichthyosiform erythroderma (NCIE). Multiple transcript variants encoding different isoforms have been found for this gene
Sequence: MAVYRLCVTTGPYLRAGTLDNISVTLVGTCGESPKQRLDRMGRDFAPGSVQKYKVRCTAELGELLLLRVHKERYAFFRKDSWYCSRICVTEPDGSVSHFPCYQWIEGYCTVELRPGTARTICQDSLPLLLDHRTRELRARQECYRWKIYAPGFPCMVDVNSFQEMESDKKFALTKTTTCVDQGDSSGNRYLPGFPMKIDIPSLMYMEPNVRYSATKTISLLFNAIPASLGMKLRGLLDRKGSWKKLDDMQNIFWCHKTFTTKYVTEHWCEDHFFGYQYLNGVNPVMLHCISSLPSKLPVTNDMVAPLLGQDTCLQTELERGNIFLADYWILAEAPTHCLNGRQQYVAAPLCLLWLSPQGALVPLAIQLSQTPGPDSPIFLPTDSEWDWLLAKTWVRNSEFLVHENNTHFLCTHLLCEAFAMATLRQLPLCHPIYKLLLPHTRYTLQVNTIARATLLNPEGLVDQVTSIGRQGLIYLMSTGLAHFTYTNFCLPDSLRARGVLAIPNYHYRDDGLKIWAAIESFVSEIVGYYYPSDASVQQDSELQAWTGEIFAQAFLGRESSGFPSRLCTPGEMVKFLTAIIFNCSAQHAAVNSGQHDFGAWMPNAPSSMRQPPPQTKGTTTLKTYLDTLPEVNISCNNLLLFWLVSQEPKDQRPLGTYPDEHFTEEAPRRSIAAFQSRLAQISRDIQERNQGLALPYTYLDPPLIENSVSISpecifica
ALOXE3 (Human) Recombinant Protein (P01) | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Yes | |
106.9kDa |