Learn More
Abnova™ Human APBB1 Partial ORF (NP_001155, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00000322-Q02.10ug
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
The protein encoded by this gene is a member of the Fe65 protein family. It is an adaptor protein localized in the nucleus. It interacts with the Alzheimer's disease amyloid precursor protein (APP), transcription factor CP2/LSF/LBP1 and the low-density lipoprotein receptor-related protein. APP functions as a cytosolic anchoring site that can prevent the gene product's nuclear translocation. This encoded protein could play an important role in the pathogenesis of Alzheimer's disease. It is thought to regulate transcription. Also it is observed to block cell cycle progression by downregulating thymidylate synthase expression. Multiple alternatively spliced transcript variants have been described for this gene but some of their full length sequence is not known. [provided by RefSeq]
Sequence: MSVPSSLSQSAINANSHGGPALSLPLPLHAAHNQLLNAKLQATAVGPKDLRSAMGEGGGPEPGPANAKWLKEGQNQLRRAATAHRDQNRNVTLTLAEEASSpecifica
NP_001155 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSVPSSLSQSAINANSHGGPALSLPLPLHAAHNQLLNAKLQATAVGPKDLRSAMGEGGGPEPGPANAKWLKEGQNQLRRAATAHRDQNRNVTLTLAEEAS | |
RUO | |
APBB1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
322 | |
APBB1 (Human) Recombinant Protein (Q02) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FE65/MGC:9072/RIR | |
APBB1 | |
Recombinant | |
wheat germ expression system |