missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ARVCF Partial ORF (NP_001661, 863 a.a. - 962 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00000421-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16131931

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Armadillo Repeat gene deleted in Velo-Cardio-Facial syndrome (ARVCF) is a member of the catenin family which play an important role in the formation of adherens junction complexes, which are thought to facilitate communication between the inside and outside environments of a cell. ARVCF gene was isolated in the search for the genetic defect responsible for the autosomal dominant Velo-Cardio-Facial syndrome (VCFS) a relatively common human disorder with phenotypic features including cleft palate, conotruncal heart defects and facial dysmorphology. ARVCF gene encodes a protein containing two motifs, a coiled coil domain in the N-terminus and a 10 armadillo repeat sequence in the midregion. Since these sequences can facilitate protein-protein interactions ARVCF is thought to function in a protein complex. In addition, ARVCF contains a predicted nuclear-targeting sequence suggesting that it may have a function as a nuclear protein. [provided by RefSeq]

Sequence: LSPGGFDDSTLPLVDKSLEGEKTGSRDVIPMDALGPDGYSTVDRRERRPRGASSAGEASEKEPLKLDPSRKAPPPGPSRPAVRLVDAVGDAKPQPVDSWV
Specifica

Specifica

NP_001661
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LSPGGFDDSTLPLVDKSLEGEKTGSRDVIPMDALGPDGYSTVDRRERRPRGASSAGEASEKEPLKLDPSRKAPPPGPSRPAVRLVDAVGDAKPQPVDSWV
RUO
ARVCF
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
421
ARVCF (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ35345
ARVCF
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali