Learn More
R&D Systems™ Human beta-Arrestin 2 (NP_004304.1) VersaClone cDNA
Specifica
Specifica
| Gene | beta-Arrestin 2 |
| Numero di accesso | NP_004304.1 |
| ID gene (immissione) | 409 |
| Alias gene | ARB2, ARR2, arrestin 3, Arrestin beta-2, arrestin, beta 2, BARR2, beta-arrestin-2, DKFZp686L0365 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantità | 10 μg |