missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BMP5 Partial ORF (NP_066551.1, 323 a.a. - 454 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00000653-Q03.25ug

Dettagli aggiuntivi : Peso : 0.02000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16063675

  • 458.00€ / 25µg


Spedizione stimata
26-10-2020
Aggiungi al carrello

Descrizione

Descrizione

Sequence: NQNRNKSSSHQDSSRMSSVGDYNTSEQKQACKKHELYVSFRDLGWQDWIIAPEGYAAFYCDGECSFPLNAHMNATNHAIVQTLVHLMFPDHVPKPCCAPTKLNAISVLYFDDSSNVILKKYRNMVVRSCGCH
Specifica

Specifica

NP_066551.1
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
40.15kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NQNRNKSSSHQDSSRMSSVGDYNTSEQKQACKKHELYVSFRDLGWQDWIIAPEGYAAFYCDGECSFPLNAHMNATNHAIVQTLVHLMFPDHVPKPCCAPTKLNAISVLYFDDSSNVILKKYRNMVVRSCGCH
RUO
BMP5
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
653
BMP5 (Human) Recombinant Protein (Q03)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC34244
BMP5
Yes
Videos
Documenti