Learn More
Abcam Human BRAP knockout HEK-293T cell line
Codice prodotto. 32142067 Sfoglia Tutto Abcam Prodotti
Cambia vista
Click to view available options
Quantità:
2 x 1,000,000 Cells
Dimensione della confezione:
pezzi 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
32142067 2 x 1,000,000 Cells pezzi 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 32142067 Fornitore Abcam N. del fornitore ab2672362x1000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 16-11-2026
Aggiungi al carrello
Aggiungi al carrello
missing translation for 'validMainframeMsg'
BRAP KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp deletion in exon 1 and 1 bp insertion in exon 1 and 8 bp deletion in exon 1. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/v...
BRAP KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp deletion in exon 1 and 1 bp insertion in exon 1 and 8 bp deletion in exon 1. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Quantità 2 x 1,000,000 Cells