missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BRPF1 Partial ORF (NP_001003694, 500 a.a. - 590 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00007862-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16128025

  • 508.00€ / 25µg
Spedizione stimata: 28-08-2024
per visualizzare lo stock disponibile

Descrizione

Descrizione

The protein encoded by this gene is expressed ubiquitously and at the highest level in testes and spermatogonia. The protein is localized within nuclei, and it is very similar in structure to two zinc finger proteins, AF10 and AF17. It is suggested that these proteins form a family of regulatory proteins. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: ILAEKRAAAPVVSVPCIPPHRLSKITNRLTIQRKSQFMQRLHSYWTLKRQSRNGVPLLRRLQTHLQSQRNCDQVGRDSEDKNWALKEQLKS
Specifica

Specifica

NP_001003694
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.75kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ILAEKRAAAPVVSVPCIPPHRLSKITNRLTIQRKSQFMQRLHSYWTLKRQSRNGVPLLRRLQTHLQSQRNCDQVGRDSEDKNWALKEQLKS
RUO
BRPF1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
7862
BRPF1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BR140
BRPF1
Recombinant
wheat germ expression system
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali