missing translation for 'onlineSavingsMsg'
Learn More

Human Carbonic Anhydrase IX/CA9 Antibody, R&D Systems™
GREENER_CHOICE

Codice prodotto. 18781717
Cambia vista
Click to view available options
Quantità:
100 μg
25 μg
500 μg
Dimensione della confezione:
100µg
25µg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18781717 25 μg 25µg
18771099 100 μg 100µg
18751137 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18781717 Fornitore R&D Systems N. del fornitore MAB2188SP

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody has been used in 3 publications

Carbonic Anhydrase IX/CA9 Monoclonal specifically detects Carbonic Anhydrase IX/CA9 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifica

Antigene Carbonic Anhydrase IX/CA9
Applicazioni Flow Cytometry, Immunoprecipitation, CyTOF, ELISA
Classificazione Monoclonal
Clone 303123
Coniugato Unconjugated
Diluizione Flow Cytometry 0.25 ug/10^6 cells, Immunoprecipitation 25 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulazione Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N. accesso geni Q16790
Alias gene CA9, CAIX, CA-IX, Carbonate dehydratase IX, carbonic anhydrase 9, carbonic anhydrase IX, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MN, P54/58N, PMW1, RCC, RCC-associated antigen G250, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250
Simboli geni CA9
Specie ospite Mouse
Immunogeno Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase IX Pro59-Asp414 Accession # Q16790
Metodo di purificazione Protein A or G purified from hybridoma culture supernatant
Quantità 25 μg
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 768
Test di specificità Detects human Carbonic Anhydrase IX (CA9) in direct ELISAs. In direct ELISAs, this antibody does not cross-react with recombinant mouse (rm) CA9 or with rhCA1, 2, 3, 4, 5A, 6, 7, 8, 10, 12, 13, or 14.
Ricostituzione Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Forma Purified
Isotype IgG2a
Vedi altri risultati Mostra meno risultati

For Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.