Learn More
Human CCL21/6Ckine Antibody, R&D Systems™
Codice prodotto. 18078671
Cambia vista
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18078671 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18078671 Fornitore R&D Systems N. del fornitore AF366
Goat Polyclonal Antibody has been used in 15 publications
CCL21/6Ckine Polyclonal specifically detects CCL21/6Ckine in Human samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifica
Antigene CCL21/6Ckine
Applicazioni Western Blot, Neutralization, Immunocytochemistry
Classificazione Polyclonal
Concentrazione LYOPH
Coniugato Unconjugated
Diluizione Western Blot 0.1 ug/mL, Neutralization 1-4 ug/mL, Immunocytochemistry 5-15 ug/mL
N. accesso geni Q6ICR7
Alias gene 6Ckine, 6CkineSmall-inducible cytokine A21, Beta-chemokine exodus-2, chemokine (C-C motif) ligand 21, CKb9, exodus-2, member 21, SCYA21, SCYA21MGC34555, secondary lymphoid tissue chemokine, SLC, SLCSecondary lymphoid-tissue chemokine, TCA-4
Simboli geni CCL21
Specie ospite Goat
Immunogeno E. coli-derived recombinant human CCL21/6Ckine Ser24-Pro134 Accession # Q6ICR7
Metodo di purificazione Affinity Purified
Quantità 100 μg
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 6366
Test di specificità Detects human CCL21/6Ckine in direct ELISAs and Western blots.
Ricostituzione Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG