Learn More
Abcam Human CCNT1 knockout HeLa cell line
Codice prodotto. 32156572 Sfoglia Tutto Abcam Prodotti
Codice prodotto. 32156572 Fornitore Abcam N. del fornitore ab2625241000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 03-11-2026
Aggiungi al carrello
Aggiungi al carrello
missing translation for 'validMainframeMsg'
CCNT1 KO cell line available to order. KO validated by Next Generation Sequencing. Free of charge wild type control available. 13 bp deletion after Ile71 of the WT protein. Knockout. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knockout cells:...
CCNT1 KO cell line available to order. KO validated by Next Generation Sequencing. Free of charge wild type control available. 13 bp deletion after Ile71 of the WT protein. Knockout. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement