missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CD96 His-tag, Recombinant Protein
HEK293 cells Recombinant Protein
Marca: Invitrogen™ A42595
Dettagli aggiuntivi : Peso : 0.01820kg
Descrizione
CD96 is a cell adhesion receptor also known as TACTILE. A member of the Ig superfamily, CD96 is a single pass transmembrane protein consisting of three extracellular Ig-like domains and a proline/serine/threonine rich region. CD96 is expressed on resident monocytes and T cells at all developmental stages (including gamma-delta T cells, thymocytes, and NK cells). In the mouse, this receptor is not expressed on B cells. Unlike in humans, mouse CD96 does not exist as alternative splice variants. Reports indicate that CD96 binds CD155/poliovirus receptor and Nectin-1 to mediate NK cell adhesion to target cells and cytotoxicity.Specifica
P40200 | |
PBS with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | |
Human CD96 His-tag | |
HEK293 cells | |
RUO | |
1700109I12Rik; CD96; CD96 antigen; CD96 molecule; cell surface antigen CD96; DKFZp667E2122; sCD96; soluble CD96; T cell activation, increased late expression; t cell-activated increased late expression protein; TACTILE; T-cell surface protein tactile | |
Human | |
Recombinant | |
≥ 85% |
Control, ELISA, Immunohistochemistry, Western Blot | |
10225 | |
5 x 50 μg | |
-20°C | |
<1 EU/μg | |
Cd96 | |
Unconjugated | |
Lyophilized |