missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CDH12 Partial ORF (NP_004052.2, 372 a.a. - 470 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00001010-Q01.25ug

Dettagli aggiuntivi : Peso : 0.02000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16053815

  • 467.00€ / 25µg


Spedizione stimata
13-01-2021
Aggiungi al carrello

Descrizione

Descrizione

Sequence: KISVLDVDEPPVFSKPLYTMEVYEDTPVGTIIGAVTAQDLDVGSSAVRYFIDWKSDGDSYFTIDGNEGTIATNELLDRESTAQYNFSIIASKVSNPLLT
Specifica

Specifica

NP_004052.2
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KISVLDVDEPPVFSKPLYTMEVYEDTPVGTIIGAVTAQDLDVGSSAVRYFIDWKSDGDSYFTIDGNEGTIATNELLDRESTAQYNFSIIASKVSNPLLT
RUO
CDH12
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
1010
CDH12 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CDHB|FLJ34857
CDH12
Yes
Videos
SDS
Documenti