missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CDK7 Partial ORF (AAH00834, 1 a.a. - 130 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00001022-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16163031

  • 335.00€ / 10µg
Spedizione stimata: 30-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The protein encoded by this gene is a member of the cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This protein forms a trimeric complex with cyclin H and MAT1, which functions as a Cdk-activating kinase (CAK). It is an essential component of the transcription factor TFIIH, that is involved in transcription initiation and DNA repair. This protein is thought to serve as a direct link between the regulation of transcription and the cell cycle. [provided by RefSeq]

Sequence: MALDVKSRAKRYEKLDFLGEGQFATVYKARDKNTNQIVAIKKIKLGHRSEAKDGINRTALREIKLLQELSHPNIIGLLDAFGHKSNISLVFDFMETDLEVIIKDNSLVLTPSHIKAYMLMTLQGLEYLHQ
Specifica

Specifica

AAH00834
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
39.71kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MALDVKSRAKRYEKLDFLGEGQFATVYKARDKNTNQIVAIKKIKLGHRSEAKDGINRTALREIKLLQELSHPNIIGLLDAFGHKSNISLVFDFMETDLEVIIKDNSLVLTPSHIKAYMLMTLQGLEYLHQ
RUO
CDK7
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
1022
CDK7 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAK1/CDKN7/MO15/STK1/p39MO15
CDK7
Recombinant
wheat germ expression system
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali