missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CLDN7 Partial ORF (NP_001298, 31 a.a. - 81 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00001366-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16126604

  • 528.00€ / 25µg
Spedizione stimata: 03-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Claudins, such as CLDN7, are involved in the formation of tight junctions between epithelial cells. Tight junctions restrict lateral diffusion of lipids and membrane proteins, and thereby physically define the border between the apical and basolateral compartments of epithelial cells (Zheng et al., 2003 [PubMed 14502431]).[supplied by OMIM]

Sequence: QMSSYAGDNIITAQAMYKGLWMDCVTQSTGMMSCKMYDSVLALSAALQATR
Specifica

Specifica

NP_001298
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.35kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QMSSYAGDNIITAQAMYKGLWMDCVTQSTGMMSCKMYDSVLALSAALQATR
RUO
CLDN7
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
1366
CLDN7 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CEPTRL2/CPETRL2/Hs.84359/claudin-1
CLDN7
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali