missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CNTF (NP_000605) VersaClone cDNA
Descrizione
- Also known as: HCNTF
- CNTF is a polypeptide hormone whose actions appear to be restricted to the nervous system where it promotes neurotransmitter synthesis and neurite outgrowth in certain neuronal populations.
- CNTF is a potent survival factor for neurons and oligodendrocytes and may be relevant in reducing tissue destruction during inflammatory attacks.
- A mutation in CNTF, which results in aberrant splicing, leads to ciliary neurotrophic factor deficiency, but this phenotype is not causally related to neurologic disease.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifica
Specifica
| Gene | hCNTF |
| Numero di accesso | NP_000605 |
| Concentrazione | 10 μg at 0.2 μg/μL |
| Formulazione | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Tipo di inserto | 616 bp |
| ID gene (immissione) | 1270 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Specie | Human |
| Quantità | 10 μg |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?